Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB545 Rat Anti-Mouse Ly-76 (TER-119), BD Horizon™
Rat Monoclonal Antibody
$162.00 - $332.00
Specifications
Antigen | Ly-76 (TER-119) |
---|---|
Clone | TER-119 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Ly-76 (TER-119) Monoclonal antibody specifically detects Ly-76 (TER-119) in Mouse samples. It is validated for Flow cytometrySpecifications
Ly-76 (TER-119) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
104231 | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
TER-119 | |
Flow Cytometry | |
RB545 | |
Rat | |
Mouse | |
Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119 | |
Mouse Fetal Liver | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title