Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Hamster Anti-Mouse CD183 (CXCR3), BD OptiBuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 757290
Description
CD183 (CXCR3) Monoclonal antibody specifically detects CD183 (CXCR3) in Mouse samples. It is validated for Flow cytometry
Specifications
CD183 (CXCR3) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
CXCR3-173 | |
RB705 | |
CXCR3; Cxcr3; CXC-R3; CXCR-3; GPR9; IP-10 Receptor | |
Mouse CXCR3 peptide (amino acids 1–37) Peptide | |
50 μg | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction