Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB705 Hamster Anti-Mouse CD3e, BD Horizon™

Catalog No. BDB570648 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570648 25 μg
BDB570560 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570648 Supplier BD Biosciences Supplier No. 570648
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

CD3e Monoclonal antibody specifically detects CD3e in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3e
Applications Flow Cytometry
Classification Monoclonal
Clone 145-2C11
Concentration 0.2 mg/mL
Conjugate RB705
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD3; CD3 epsilon; Cd3e; CD3ε; T3e
Host Species Hamster
Immunogen H-2Kb specific cytotoxic T lymphocyte clone BM10-37
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Gene ID (Entrez) 12501
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.