Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Hamster Anti-Mouse Fc ε Receptor Iα (FcεR1α), BD OptiBuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 756904
Description
Fc ε Receptor Iα (FcεR1α) Monoclonal antibody specifically detects Fc ε Receptor Iα (FcεR1α) in Mouse samples. It is validated for Flow cytometry
Specifications
Fc ε Receptor Iα (FcεR1α) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
MAR-1 (also known as MAR1) | |
RB705 | |
FceR1 alpha; Fcer1a; FcεRIα; High affinity IgE Receptor; MAR1 | |
Mouse FcεRIα/β/γ-transfected CHO cells | |
50 μg | |
14125 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction