Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Hamster Anti-Mouse TCR Vβ3, BD OptiBuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 757958
Description
TCR Vβ3 Monoclonal antibody specifically detects TCR Vβ3 in Mouse samples. It is validated for Flow cytometry
Specifications
TCR Vβ3 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
KJ25 | |
RB705 | |
TCR V beta 3; TCR Vβ3 | |
αβ TCR purified from mouse T-cell hybridoma 2B4.6 | |
50 μg | |
100039790 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction