Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD11a (LFA-1A), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570270
Description
CD11a (LFA-1A) Monoclonal antibody specifically detects CD11a (LFA-1A) in Human samples. It is validated for Flow cytometry
Specifications
CD11a (LFA-1A) | |
Monoclonal | |
RB705 | |
LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
HI111 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
3683 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction