Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD127 (IL-7 Receptor α), BD Horizon™
Mouse Monoclonal Antibody
$175.50 - $447.50
Specifications
| Antigen | CD127 (IL-7 Receptor α) |
|---|---|
| Clone | HIL-7R-M21 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | RB705 |
Description
CD127 (IL-7 Receptor α) Monoclonal antibody specifically detects CD127 (IL-7 Receptor α) in Human samples. It is validated for Flow cytometrySpecifications
| CD127 (IL-7 Receptor α) | |
| Flow Cytometry | |
| RB705 | |
| Mouse | |
| Human | |
| IL-7R; IL7R; IL7RA; IL-7Rα; IL-7R-alpha; Interleukin-7 Receptor alpha | |
| Human IL-7R Recombinant Protein | |
| Affinity Chromatography |
| HIL-7R-M21 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 3575 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title