Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD147 (EMMPRIN), BD Horizon™
Mouse Monoclonal Antibody
$166.00 - $324.00
Specifications
Antigen | CD147 (EMMPRIN) |
---|---|
Clone | HIM6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB705 |
Description
CD147 (EMMPRIN) Monoclonal antibody specifically detects CD147 (EMMPRIN) in Human samples. It is validated for Flow cytometrySpecifications
CD147 (EMMPRIN) | |
Flow Cytometry | |
RB705 | |
Mouse | |
Human | |
BSG; Basigin; BASI; Neurothelin; 5F7; EMMPRIN; M6; OK; TCSF | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HIM6 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
682 | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title