Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD152 (CTLA-4), BD Horizon™
Mouse Monoclonal Antibody
$150.00 - $376.00
Specifications
Antigen | CD152 (CTLA-4) |
---|---|
Clone | BNI3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB705 |
Description
CD152 (CTLA-4) Monoclonal antibody specifically detects CD152 (CTLA-4) in Human samples. It is validated for Flow cytometry|Intracellular staining (flow cytometry)Specifications
CD152 (CTLA-4) | |
Flow Cytometry | |
RB705 | |
Mouse | |
Human | |
CTLA-4; AILIM; Cytotoxic T-lymphocyte protein 4 | |
Human CTLA4 Recombinant Protein | |
Affinity Chromatography |
BNI3 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
1493 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title