Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
RB705 Mouse Anti-Human CD183 (CXCR3), BD Horizon™
Mouse Monoclonal Antibody
$160.00 - $406.00
Specifications
| Antigen | CD183 (CXCR3) | 
|---|---|
| Clone | 1C6/CXCR3 (also known as 1C6, LS177-1C6) | 
| Applications | Flow Cytometry | 
| Classification | Monoclonal | 
| Conjugate | RB705 | 
Description
CD183 (CXCR3) Monoclonal antibody specifically detects CD183 (CXCR3) in Human samples. It is validated for Flow cytometrySpecifications
| CD183 (CXCR3) | |
| Flow Cytometry | |
| RB705 | |
| Mouse | |
| Human | |
| CXCR3; C-X-C chemokine receptor type 3; GPR9; IP10R; MigR; CKRL2; CMKAR3 | |
| Human CXCR3 Peptide | |
| Affinity Chromatography | 
| 1C6/CXCR3 (also known as 1C6, LS177-1C6) | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 2833 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
             
                                                