Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD19, BD Horizon™
Mouse Monoclonal Antibody
$152.00 - $362.00
Specifications
Antigen | CD19 |
---|---|
Clone | SJ25C1 (also known as SJ25-C1) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB705 |
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow cytometrySpecifications
CD19 | |
Flow Cytometry | |
RB705 | |
Mouse | |
Human | |
B4; B-lymphocyte antigen CD19; Leu-12 | |
Human NALM1 + NALM16 cells | |
Affinity Chromatography |
SJ25C1 (also known as SJ25-C1) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
930 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title