Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD192 (CCR2), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 757633
Description
CD192 (CCR2) Monoclonal antibody specifically detects CD192 (CCR2) in Human samples. It is validated for Flow cytometry
Specifications
CD192 (CCR2) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
LS132.1D9 (also known as 1D9) | |
RB705 | |
CCR2; CD192; CKR2; CC-CKR-2; CMKBR2; MCP-1 receptor; MCP-1-R | |
Human CCR2 Transfected Cell Line | |
50 μg | |
729230 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction