Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD20, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570660
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow cytometry
Specifications
CD20 | |
Monoclonal | |
RB705 | |
MS4A1; B1; Bp35; LEU-16; S7 | |
Human 6.16c1.3 B cell line | |
25 Tests | |
931 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2b κ |
Flow Cytometry | |
2H7 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction