Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD279 (PD-1), BD Horizon™
Mouse Monoclonal Antibody
$160.00 - $396.00
Specifications
Antigen | CD279 (PD-1) |
---|---|
Clone | EH12.1 (also known as EH12) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB705 |
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow cytometrySpecifications
CD279 (PD-1) | |
Flow Cytometry | |
RB705 | |
Mouse | |
Human | |
PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2; hSLE1 | |
Human PD-1 Recombinant Protein | |
Affinity Chromatography |
EH12.1 (also known as EH12) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
5133 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title