Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD69, BD Horizon™
Mouse Monoclonal Antibody
$168.00 - $412.00
Specifications
Antigen | CD69 |
---|---|
Clone | FN50 (also known as FN 50) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB705 |
Description
CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Flow cytometrySpecifications
CD69 | |
Flow Cytometry | |
RB705 | |
Mouse | |
Human | |
AIM; CLEC2C; EA1; GP32/28; Leu23; MLR-3; VEA; BL-AC/P26 | |
Anti-μ stimulated human B lymphocytes | |
Affinity Chromatography |
FN50 (also known as FN 50) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
969 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title