Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD81 (TAPA-1), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 757589
Description
CD81 (TAPA-1) Monoclonal antibody specifically detects CD81 (TAPA-1). It is validated for Flow cytometry
Specifications
| CD81 (TAPA-1) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| 975 | |
| Liquid |
| Flow Cytometry | |
| JS-81 (also known as JS81) | |
| RB705 | |
| TAPA1; Tetraspanin-28; Tspan-28; TSPAN28; CVID6; S5.7; M38 | |
| Affinity Chromatography | |
| RUO | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction