Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD8b, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 756931
Description
CD8b Monoclonal antibody specifically detects CD8b in Human samples. It is validated for Flow cytometry
Specifications
CD8b | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
2ST8.5H7 (also known as T8/2ST8-5H7) | |
RB705 | |
CD8B; CD8B1; CD8 beta; CD8β; Leu2; Ly3; LYT3 | |
CD8+ suppressor T-cell | |
50 μg | |
926 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction