Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human FoxP3, BD Horizon™
Mouse Monoclonal Antibody
$178.00 - $438.00
Specifications
Antigen | FoxP3 |
---|---|
Clone | 259D/C7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB705 |
Description
FoxP3 Monoclonal antibody specifically detects FoxP3 in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
FoxP3 | |
Flow Cytometry | |
RB705 | |
Mouse | |
Human | |
scurfin; IPEX; JM2; AIID; XPID; DIETER; PIDX | |
IgG1 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
259D/C7 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
FoxP3 | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title