Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human Granzyme B, BD Horizon™
Mouse Monoclonal Antibody
$208.00 - $469.00
Specifications
| Antigen | Granzyme B |
|---|---|
| Clone | GB11 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | RB705 |
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
| Granzyme B | |
| Flow Cytometry | |
| RB705 | |
| Mouse | |
| Human | |
| GZMB; Granzyme-2; CCPI; CGL1; CSPB; CTLA1; CTSGL1; GRB; HLP; SECT | |
| Human Granzyme B | |
| Affinity Chromatography |
| GB11 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 3002 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title