Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human IFN-γ, BD Horizon™
Mouse Monoclonal Antibody
$168.00 - $412.00
Specifications
Antigen | IFN-γ |
---|---|
Clone | B27 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB705 |
Description
IFN-γ Monoclonal antibody specifically detects IFN-γ in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
IFN-γ | |
Flow Cytometry | |
RB705 | |
Mouse | |
Human | |
IFNG; Interferon-gamma; Interferon-γ; Type II interferon; MAF | |
Human IFN-γ Recombinant Protein | |
Affinity Chromatography |
B27 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
3458 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title