Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB705 Mouse Anti-Human TCRαβ, BD Horizon™

Catalog No. BDB570702 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570702 25 Tests
BDB570614 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570702 Supplier BD Biosciences Supplier No. 570702
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCRαβ Monoclonal antibody specifically detects TCRαβ in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCRαβ
Applications Flow Cytometry
Classification Monoclonal
Clone IP26 (also known as IP26A)
Conjugate RB705
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias alpha-beta T cell receptor, TCRAB, TRA@/TRB@; IP26A
Host Species Mouse
Immunogen Human T Cell Clone
Purification Method Affinity Chromatography
Quantity 25 Tests
Regulatory Status RUO
Gene ID (Entrez) 6957, 28755
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.