Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Integrin αvβ5, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 757111
Description
Integrin αvβ5 Monoclonal antibody specifically detects Integrin αvβ5. It is validated for Flow cytometry
Specifications
Integrin αvβ5 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2b κ |
Flow Cytometry | |
ALULA | |
RB705 | |
Integrin alpha v beta 5; αVβ5; CD51/β5 | |
Mouse fibroblasts expressing αvβ5 | |
50 μg | |
3685, 3693, 16410, 16419 | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction