Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Mouse CD161b,c (NK-1.1), BD Horizon™
Mouse Monoclonal Antibody
$104.00 - $350.00
Specifications
Antigen | CD161b,c (NK-1.1) |
---|---|
Clone | PK136 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD161b,c (NK-1.1) Monoclonal antibody specifically detects CD161b,c (NK-1.1) in Mouse samples. It is validated for Flow cytometrySpecifications
CD161b,c (NK-1.1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
17059, 80782 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
PK136 | |
Flow Cytometry | |
RB705 | |
Mouse | |
Mouse | |
Klrb1b, CD161b, Nkrp1b; Klrb1c, CD161c, NK1.1, Nkrp1c | |
Mouse NK-1+ Spleen and Bone Marrow Cells | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title