Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB705 Mouse Anti-Rat CD90/Mouse CD90.1, BD OptiBuild™

Catalog No. BDB756990 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB756990 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB756990 Supplier BD Biosciences Supplier No. 756990
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD90/CD90.1 Monoclonal antibody specifically detects CD90/CD90.1. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD90/CD90.1
Applications Flow Cytometry
Classification Monoclonal
Clone OX-7 (also known as OX7)
Concentration 0.2 mg/mL
Conjugate RB705
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Rat Thy-1; Mouse Thy-1.1
Host Species Mouse
Immunogen Rat Thymocyte Thy-1 Antigen
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Gene ID (Entrez) 21838, 24832
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.