Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-TCF7 (TCF1), BD Horizon™
Mouse Monoclonal Antibody
$137.00 - $347.00
Specifications
| Antigen | TCF7 (TCF1) |
|---|---|
| Clone | S33-966 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
TCF7 (TCF1) Monoclonal antibody specifically detects TCF7 (TCF1) in Mouse|Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
| TCF7 (TCF1) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 6932, 21414 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| S33-966 | |
| Flow Cytometry | |
| RB705 | |
| Mouse | |
| Mouse, Human | |
| TCF-1; TCF-7; Tcf1; Tcf7; transcription factor 7 | |
| Mouse TCF-7 Recombinant Protein | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title