Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD117 (c-Kit), BD Horizon™
Rat Monoclonal Antibody
$130.00 - $350.00
Specifications
Antigen | CD117 (c-Kit) |
---|---|
Clone | 2B8 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD117 (c-Kit) Monoclonal antibody specifically detects CD117 (c-Kit) in Mouse samples. It is validated for Flow cytometrySpecifications
CD117 (c-Kit) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
16590 | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
2B8 | |
Flow Cytometry | |
RB705 | |
Rat | |
Mouse | |
c-KIT; W; SCFR; Stem Cell Factor Receptor; Sl; Steel Factor Receptor; Ssm | |
Mouse Bone Marrow Mast Cells | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title