Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD147, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756943
Description
CD147 Monoclonal antibody specifically detects CD147 in Mouse samples. It is validated for Flow cytometry
Specifications
CD147 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
RL73 (also known as RL73.2) | |
RB705 | |
Bsg; Basigin; BASI; EMMPRIN; gp 42; HT7; HT-7; Neurothelin | |
Mouse EL4 Cell Subline | |
50 μg | |
12215 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction