Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD19, BD Horizon™
Rat Monoclonal Antibody
$104.00 - $334.00
Specifications
| Antigen | CD19 |
|---|---|
| Clone | 1D3 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD19 Monoclonal antibody specifically detects CD19 in Mouse samples. It is validated for Flow cytometrySpecifications
| CD19 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 12478 | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 1D3 | |
| Flow Cytometry | |
| RB705 | |
| Rat | |
| Mouse | |
| Cd19; CD19 antigen; B-lymphocyte antigen CD19 | |
| Mouse CD19 Transfected Cell Line | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title