Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD24, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756900
Description
CD24 Monoclonal antibody specifically detects CD24 in Mouse samples. It is validated for Flow cytometry
Specifications
CD24 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
30-F1 | |
RB705 | |
Cd24a; HSA; heat stable antigen; Ly-52; nectadrin | |
Mouse thymus or spleen | |
50 μg | |
12484 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction