Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD26, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 757373
Description
CD26 Monoclonal antibody specifically detects CD26 in Mouse samples. It is validated for Flow cytometry
Specifications
CD26 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
H194-112 | |
RB705 | |
Cd26; Dpp4; dipeptidylpeptidase 4; THAM, DPP IV | |
Cell-surface molecules solubilized from BALB/c mouse thymocytes | |
50 μg | |
13482 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction