Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD273 (PD-L2), BD Horizon™
Rat Monoclonal Antibody
$154.00 - $380.00
Specifications
Antigen | CD273 (PD-L2) |
---|---|
Clone | MIH37 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD273 (PD-L2) Monoclonal antibody specifically detects CD273 (PD-L2) in Mouse samples. It is validated for Flow cytometrySpecifications
CD273 (PD-L2) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
58205 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MIH37 | |
Flow Cytometry | |
RB705 | |
Rat | |
Mouse | |
B7-DC; Btdc; PD-1 ligand 2; PD-L2; PDCD1 ligand 2; programmed cell death 1 ligand 2 | |
Mouse B7-DC-transfected Cell Line | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title