Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD279 (PD-1), BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 570654
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow cytometry
Specifications
CD279 (PD-1) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
29F.1A12 | |
RB705 | |
PD-1; Pdc1; Pdcd1; mPD-1; programmed cell death 1 protein | |
PD-1 cDNA and PD-1-Ig Fusion Protein | |
25 μg | |
18566 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction