Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD309 (VEGFR2), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756852
Description
CD309 (VEGFR2) Monoclonal antibody specifically detects CD309 (VEGFR2) in Mouse samples. It is validated for Flow cytometry
Specifications
CD309 (VEGFR2) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
Avas 12α1 | |
RB705 | |
CD309; Fetal liver kinase 1; Kdr; VEGF Receptor-2; VEGFR-2; AVAS12; | |
Mouse Flk-1 Recombinant Protein | |
50 μg | |
16542 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction