Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD62L (L-Selectin), BD Horizon™
Rat Monoclonal Antibody
$142.00 - $352.00
Specifications
Antigen | CD62L (L-Selectin) |
---|---|
Clone | MEL-14 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD62L (L-Selectin) Monoclonal antibody specifically detects CD62L (L-Selectin) in Mouse samples. It is validated for Flow cytometrySpecifications
CD62L (L-Selectin) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
20343 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MEL-14 | |
Flow Cytometry | |
RB705 | |
Rat | |
Mouse | |
Sell; L-selectin; LECAM-1; LAM-1; Lnhr; Ly-22; Ly-m22; Lyam-1 | |
C3H/eb mouse B lymphoma 38C-13 | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title