Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse CD62P (P-Selectin), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756824
Description
CD62P (P-Selectin) Monoclonal antibody specifically detects CD62P (P-Selectin) in Mouse samples. It is validated for Flow cytometry
Specifications
CD62P (P-Selectin) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
RB40.34 | |
RB705 | |
P-selectin; Selp; LECAM3; LYAM3; PADGEM; GMP-140; Grmp | |
P-selectin-IgG1 Fusion | |
50 μg | |
20344 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction