Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB705 Rat Anti-Mouse CD90.2, BD OptiBuild™

Catalog No. BDB757459 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB757459 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB757459 Supplier BD Biosciences Supplier No. 757459
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD90.2 Monoclonal antibody specifically detects CD90.2 in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD90.2
Applications Flow Cytometry
Classification Monoclonal
Clone 30-H12
Concentration 0.2 mg/mL
Conjugate RB705
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Thy-1.2; T25; Thymus Cell Antigen 1; Theta
Host Species Rat
Immunogen Mouse Thymus / Spleen
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Gene ID (Entrez) 21838
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.