Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse Ly-49D, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 757097
Description
Ly-49D Monoclonal antibody specifically detects Ly-49D in Mouse samples. It is validated for Flow cytometry
Specifications
Ly-49D | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
4E5 | |
RB705 | |
Ly49d; Ly-49d; Klra4; Chok | |
Not Reported | |
50 μg | |
16635 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction