Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB705 Rat Anti-Mouse Ly-6A/E (Sca-1), BD Horizon™

Catalog No. BDB570679 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570679 25 μg
BDB570591 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570679 Supplier BD Biosciences Supplier No. 570679
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6A/E (Sca-1) Monoclonal antibody specifically detects Ly-6A/E (Sca-1) in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6A/E (Sca-1)
Applications Flow Cytometry
Classification Monoclonal
Clone D7
Concentration 0.2 mg/mL
Conjugate RB705
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ly-6A/E; Lymphocyte antigen 6A-2/6E-1; Ly-6A.2/Ly-6E.1; Sca-1; TAP
Host Species Rat
Immunogen IL-2-dependent mouse T-cell line CTL-L
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Gene ID (Entrez) 110454
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.