Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse Ly-6G/Ly-6C, BD Horizon™
Rat Monoclonal Antibody
$120.00 - $228.00
Specifications
Antigen | Ly-6G/Ly-6C |
---|---|
Clone | RB6-8C5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Ly-6G/Ly-6C Monoclonal antibody specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow cytometrySpecifications
Ly-6G/Ly-6C | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
17067, 546644 | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
RB6-8C5 | |
Flow Cytometry | |
RB705 | |
Rat | |
Mouse | |
Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1 | |
Not Reported | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title