Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse TCR Vα2, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756886
Description
TCR Vα2 Monoclonal antibody specifically detects TCR Vα2 in Mouse samples. It is validated for Flow cytometry
Specifications
TCR Vα2 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
B20.1 | |
RB705 | |
T cell receptor alpha, variable region 2 | |
Soluble αβ TCR from mouse cytotoxic T-cell clone KB5-C20 | |
50 μg | |
111660 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction