Learn More
Description
Specifications
Specifications
| Applications | Flow Cytometry |
| Concentration | 0.1 mg/mL |
| Conjugate | RB705 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Content And Storage | Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.