Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Hamster Anti-Mouse TCR Vγ 3, BD OptiBuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 756861
Description
TCR Vγ 3 Monoclonal antibody specifically detects TCR Vγ 3 in Mouse samples. It is validated for Flow cytometry
Specifications
TCR Vγ 3 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
536 | |
RB744 | |
TCR Vγ3; TCR V gamma 3; TCR Vg3 | |
AKR mouse dendritic epidermal cell clone 7-17 | |
50 μg | |
110067 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction