Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Hamster Anti-Mouse TCR Vδ6.3/6.2, BD OptiBuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 756821
Description
TCR Vδ6.3/6.2 Monoclonal antibody specifically detects TCR Vδ6.3/6.2 in Mouse samples. It is validated for Flow cytometry
Specifications
TCR Vδ6.3/6.2 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
8F4H7B7 | |
RB744 | |
V delta 6.3/2 TCR; TCR Vδ 6.3/2; TCR Vδ6.3/2; TCR Vd6.3/2 | |
1E6.11.D7, a C57BL/6 mouse Vγ1.1Vδ6.3 TCR-bearing IEL/T-cell hybridoma | |
50 μg | |
110066 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction