Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Hamster Anti-Rat/Mouse CD49a (Integrin α1), BD OptiBuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 756815
Description
CD49a (Integrin α1) Monoclonal antibody specifically detects CD49a (Integrin α1). It is validated for Flow cytometry
Specifications
CD49a (Integrin α1) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2 λ1 |
Flow Cytometry | |
Ha31/8 | |
RB744 | |
Itga1; Integrin alpha-1; Integrin α1; Laminin and collagen receptor; VLA-1a | |
Rat Emulsified Lewis Rat Glomeruli Cells | |
50 μg | |
25118, 109700 | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction