Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human ACT β2 Itgrn (CD18), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 757570
Description
ACT β2 Itgrn (CD18) Monoclonal antibody specifically detects ACT β2 Itgrn (CD18) in Human samples. It is validated for Flow cytometry
Specifications
ACT β2 Itgrn (CD18) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
mAb 24 (also known as 24; m24; M24; mAb24) | |
RB744 | |
ITGB2; Integrin beta-2; Integrin β2; LFA-1 β; LFA1b; beta 2 integrin; β2 integrin | |
Fibronectin-purified Human Monocytes | |
50 μg | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction