Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human C3a Receptor, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 756885
Description
C3a Receptor Monoclonal antibody specifically detects C3a Receptor in Human samples. It is validated for Flow cytometry
Specifications
C3a Receptor | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2b |
Flow Cytometry | |
hC3aRZ8 | |
RB744 | |
C3AR; C3a-R; C3AR1; C3a anaphylatoxin chemotactic receptor; AZ3B | |
Human C3aR Transfected Cell Line | |
50 μg | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction