Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human CD105 (Endoglin), BD Horizon™
Mouse Monoclonal Antibody
$160.00 - $406.00
Specifications
Antigen | CD105 (Endoglin) |
---|---|
Clone | 266 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB744 |
Description
CD105 (Endoglin) Monoclonal antibody specifically detects CD105 (Endoglin) in Human samples. It is validated for Flow cytometrySpecifications
CD105 (Endoglin) | |
Flow Cytometry | |
RB744 | |
Mouse | |
Human | |
END; Endoglin; HHT1; MSC; ORW; ORW1 | |
Human Umbilical Vein Endothelial Cells | |
Affinity Chromatography |
266 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
2022 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title