Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human CD11a (LFA-1A), BD Horizon™
Mouse Monoclonal Antibody
$152.00 - $376.00
Specifications
Antigen | CD11a (LFA-1A) |
---|---|
Clone | HI111 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB744 |
Description
CD11a (LFA-1A) Monoclonal antibody specifically detects CD11a (LFA-1A) in Human samples. It is validated for Flow cytometrySpecifications
CD11a (LFA-1A) | |
Flow Cytometry | |
RB744 | |
Mouse | |
Human | |
LFA-1α; Lymphocyte (Leukocyte) function-associated antigen 1 α chain; ITGAL | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HI111 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
3683 | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title