Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human CD130 (gp130), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 756845
Description
CD130 (gp130) Monoclonal antibody specifically detects CD130 (gp130) in Human samples. It is validated for Flow cytometry
Specifications
CD130 (gp130) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
AM64 | |
RB744 | |
IL6ST; IL6RB; IL-6Rβ; Interleukin-6 Receptor beta; gp130 | |
Purified Human CD130 | |
50 μg | |
3572 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction