Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human CD203c, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 757659
Description
CD203c Monoclonal antibody specifically detects CD203c in Human samples. It is validated for Flow cytometry
Specifications
CD203c | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
NP4D6 | |
RB744 | |
ENPP3; E-NPP 3; NPP3; PDNP3; PD-Ibeta; PD-1β; B10; NPPase; gp130RB13-6 | |
Human CD203c Transfected Cell Line | |
50 μg | |
5169 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction